2-Acetyl-4-bromthiophen 97%

Lieferant: Apollo Scientific
Achtung

OR5412-1G OR5412-25G OR5412-5G
APOSOR5412-1GEA 21.9 EUR
APOSOR5412-1G APOSOR5412-25G APOSOR5412-5G
2-Acetyl-4-bromthiophen 97%
2-Acetyl-4-bromthiophen
Formel: C₆H₅BrOS
Molecular Weight: 205,08 g/mol
Siedepunkt: 68…70 °C/0,2mmHg
Lagertemperatur: Kühlschrank
MDL: MFCD01566552
CAS-Nummer: 7209-11-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR