5-Chlor-2-thiophenboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Chlor-2-thienylboronsäure, 5-Chlorthiophen-2-boronsäure
OR5421-25G
OR5421-5G
APOSOR5421-25GEA
98.6
EUR
APOSOR5421-25G
APOSOR5421-5G
5-Chlor-2-thiophenboronsäure
5-Chloro-2-thiopheneboronic acid
Formel:
C₄H₄BClO₂S Molecular Weight: 162.4 g/mol Schmelzpunkt: 136…140 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00236030 CAS-Nummer: 162607-18-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...