4-Chlor-3-nitroanisol 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Chlor-4-methoxy-2-nitrobenzol
OR5423-100G
OR5423-500G
OR5423-25G
APOSOR5423-100GEA
48
EUR
APOSOR5423-100G
APOSOR5423-500G
APOSOR5423-25G
4-Chlor-3-nitroanisol 95%
4-Chlor-3-nitroanisol
Formel:
C₇H₆ClNO₃ Molecular Weight: 187.58 g/mol Schmelzpunkt: 41…45 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007077 CAS-Nummer: 10298-80-3 EINECS: 233-674-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...