5-Nitro-2-furonitril 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Cyan-5-nitrofuran
OR5432-1G
OR5432-25G
OR5432-5G
APOSOR5432-1GEA
36.4
EUR
APOSOR5432-1G
APOSOR5432-25G
APOSOR5432-5G
5-Nitro-2-furonitril 97%
5-Nitro-2-furonitril
Formel:
C₅H₂N₂O₃ Molecular Weight: 138,08 g/mol Schmelzpunkt: 58…61 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082647 CAS-Nummer: 59-82-5 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...