Methyl-4-amino-3-(benzyloxy)-5-bromobenzenecarboxylate
Lieferant: Apollo Scientific
Synonyme:
Methyl 4-amino-3-(benzyloxy)-5-bromobenzoate
OR54325-1G
OR54325-500MG
APOSOR54325-1GEA
338
EUR
APOSOR54325-1G
APOSOR54325-500MG
Methyl-4-amino-3-(benzyloxy)-5-bromobenzenecarboxylate
Methyl-4-amino-3-(benzyloxy)-5-bromobenzenecarboxylate
Formel:
C₁₅H₁₄BrNO₃ Molecular Weight: 336.19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12827830 CAS-Nummer: 881909-58-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...