Methyl-2-(benzyloxy)-5-nitrobenzenecarboxylate

Lieferant: Apollo Scientific

Synonyme: Methyl 2-(benzyloxy)-5-nitrobenzoate

OR54326-1G OR54326-500MG OR54326-5G
APOSOR54326-1GEA 161 EUR
APOSOR54326-1G APOSOR54326-500MG APOSOR54326-5G
Methyl-2-(benzyloxy)-5-nitrobenzenecarboxylate
Methyl-2-(benzyloxy)-5-nitrobenzenecarboxylate
Formel: C₁₅H₁₃NO₅
Molecular Weight: 287,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12827823
CAS-Nummer: 508211-52-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR