2-Chloro-6-methoxy-4-[2-(trimethylsilyl)ethynyl]-3-pyridinol
Lieferant: Apollo Scientific
Synonyme:
2-Chloro-3-hydroxy-6-methoxy-4-[2-(trimethylsilyl)ethynyl]pyridine
OR54328-1G
OR54328-500MG
APOSOR54328-1GEA
423
EUR
APOSOR54328-1G
APOSOR54328-500MG
2-Chloro-6-methoxy-4-[2-(trimethylsilyl)ethynyl]-3-pyridinol
2-Chloro-6-methoxy-4-[2-(trimethylsilyl)ethynyl]-3-pyridinol
Formel:
C₁₁H₁₄ClNO₂Si Molecular Weight: 255,78 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12827784 CAS-Nummer: 208519-38-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...