N-Boc-D-alaninol

Lieferant: Apollo Scientific
Achtung

Synonyme: Boc-D-Alaninol, (R)-2-(BOC-amino)-1-propanol

OR54336-100G OR54336-25G OR54336-5G
APOSOR54336-100GEA 111 EUR
APOSOR54336-100G APOSOR54336-25G APOSOR54336-5G
N-Boc-D-alaninol
N-Boc-D-alaninol
Formel: C₈H₁₇NO₃
Molecular Weight: 175,2 g/mol
Schmelzpunkt: 56…61 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00235912
CAS-Nummer: 106391-86-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR