6-Methyl-1H-indazol 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR54339-1G
OR54339-5G
APOSOR54339-1GEA
21.9
EUR
APOSOR54339-1G
APOSOR54339-5G
6-Methyl-1H-indazol 97%
6-Methyl-1H-indazole
Formel:
C₈H₈N₂ Molecular Weight: 132,1625 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07781625 CAS-Nummer: 698-24-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...