Methyl-2-amino-5-hydroxy-4-methoxybenzoate 95+%

Lieferant: Apollo Scientific
Achtung

OR54344-250MG
APOSOR54344-250MGEA 125 EUR
APOSOR54344-250MG
Methyl-2-amino-5-hydroxy-4-methoxybenzoate 95+%
Methyl-2-amino-5-hydroxy-4-methoxybenzoate
Formel: C₉H₁₁NO₄
Molecular Weight: 197,19 g/mol
Schmelzpunkt: 160 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD17168892
CAS-Nummer: 50413-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR