4-Bromo-2-methylbenzyl alcohol

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Bromo-2-methylphenyl)methanol

OR54375-1G OR54375-5G OR54375-10G
APOSOR54375-1GEA 21.9 EUR
APOSOR54375-1G APOSOR54375-5G APOSOR54375-10G
4-Bromo-2-methylbenzyl alcohol
4-Bromo-2-methylbenzyl alcohol
Formel: C₈H₉BrO
Molecular Weight: 201,06 g/mol
Schmelzpunkt: 70…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11847398
CAS-Nummer: 17100-58-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR