3-Piperazin-1-yl-1,2-benzisoxazole 95%

Lieferant: Apollo Scientific
Achtung

OR54378-250MG OR54378-1G
APOSOR54378-250MGEA 261 EUR
APOSOR54378-250MG APOSOR54378-1G
3-Piperazin-1-yl-1,2-benzisoxazole 95%
3-Piperazin-1-yl-1,2-benzisoxazole
Formel: C₁₁H₁₃N₃O
Molecular Weight: 203,25 g/mol
Schmelzpunkt: 74…76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08060968
CAS-Nummer: 87691-89-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR