2,4,5-Trihydroxybenzaldehyd 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR54382-1G
OR54382-5G
APOSOR54382-1GEA
52.2
EUR
APOSOR54382-1G
APOSOR54382-5G
2,4,5-Trihydroxybenzaldehyd 95%
2,4,5-Trihydroxybenzaldehyde
Formel:
C₇H₆O₄ Molecular Weight: 154,12 g/mol Schmelzpunkt: 223…225 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016592 CAS-Nummer: 35094-87-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...