1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine 98+%
Lieferant: Apollo Scientific
Synonyme: DOPE, 1,2-Dioleoyl-sn-glycero-3-PE, L-α-Phosphatidylethanolamine, dioleoyl, 2-Aminoethyl-(R)-2,3-bis(oleoyloxy)propyl hydrogen phosphate, 3-sn-Phosphatidylethanolamine, 1,2-dideoyl, (Z)-(2R)-3-(((2-Aminoethoxy)(hydroxy)phosphoryl)oxy)propane-1,2-diyl dioleate
OR54392-250MG OR54392-1G OR54392-5G
APOSOR54392-250MGEA 119 EUR
APOSOR54392-250MG APOSOR54392-1G APOSOR54392-5G
1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine 98+%
1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine
Formel: C₄₁H₇₈NO₈P
Molecular Weight: 744,03 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD16621009
CAS-Nummer: 4004-05-1