2-Aminoisophthalic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Aminoisophthalic Acid
OR5441-5G
OR5441-1G
OR5441-25G
APOSOR5441-5GEA
24.8
EUR
APOSOR5441-5G
APOSOR5441-1G
APOSOR5441-25G
2-Aminoisophthalic acid 98%
2-Aminoisophthalic acid
Formel:
C₈H₇NO₄ Molecular Weight: 181.15 g/mol Schmelzpunkt: 300…304 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00817808 CAS-Nummer: 39622-79-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...