2-Thiophenboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Thienylboronsäure, Thiophen-2-boronsäure
OR5442-25G
OR5442-5G
APOSOR5442-25GEA
30.6
EUR
APOSOR5442-25G
APOSOR5442-5G
2-Thiophenboronsäure
2-Thiopheneboronic acid
Formel:
C₄H₅BO₂S Molecular Weight: 127,96 g/mol Schmelzpunkt: 128…131 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00151850 CAS-Nummer: 6165-68-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...