3,5-Dimethyl-4-isoxazolboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: (dimethyl-1,2-oxazol-4-yl)boronic acid

OR5444-5G OR5444-25G
APOSOR5444-5GEA 26.1 EUR
APOSOR5444-5G APOSOR5444-25G
3,5-Dimethyl-4-isoxazolboronsäure
3,5-Dimethylisoxazole-4-boronic acid
Formel: C₅H₈BNO₃
Molecular Weight: 140,93 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD02677945
CAS-Nummer: 16114-47-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR