6-Methoxy-1,2,3,4-tetrahydronaphthalin

Lieferant: Apollo Scientific

OR54469-1G OR54469-5G
APOSOR54469-1GEA 66.7 EUR
APOSOR54469-1G APOSOR54469-5G
6-Methoxy-1,2,3,4-tetrahydronaphthalin
6-Methoxy-1,2,3,4-tetrahydronaphthalin
Formel: C₁₁H₁₄O
Molecular Weight: 162,23 g/mol
Siedepunkt: 131 °C
Dichte: 1,03 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001737
CAS-Nummer: 1730-48-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR