N-Methylacetoacetamid

Lieferant: Apollo Scientific

OR54480-25G OR54480-500G OR54480-100G
APOSOR54480-25GEA 24.8 EUR
APOSOR54480-25G APOSOR54480-500G APOSOR54480-100G
N-Methylacetoacetamid
N-Methylacetoacetamid
Formel: C₅H₉NO₂
Flammpunkt: 105 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00044026
CAS-Nummer: 20306-75-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR