3-Bromo-3'-chloro-1,1'-biphenyl 98%

Lieferant: Apollo Scientific
Achtung

OR54490-1G OR54490-5G OR54490-25G
APOSOR54490-1GEA 21.9 EUR
APOSOR54490-1G APOSOR54490-5G APOSOR54490-25G
3-Bromo-3'-chloro-1,1'-biphenyl 98%
3-Bromo-3'-chloro-1,1'-biphenyl
Formel: C₁₂H₈BrCl
Molecular Weight: 267,55 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06201361
CAS-Nummer: 844856-42-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR