4-Methyl-2-nitrophenylisocyanat 97%

Lieferant: Apollo Scientific
Gefahr

OR54499-1G OR54499-5G
APOSOR54499-1GEA 348 EUR
APOSOR54499-1G APOSOR54499-5G
4-Methyl-2-nitrophenylisocyanat 97%
4-Methyl-2-nitrophenylisocyanat

When melted, may contain small amount of sediment.

Formel: C₈H₆N₂O₃
Molecular Weight: 178,15 g/mol
Schmelzpunkt: 56…58 °C
Flammpunkt: 113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014705
CAS-Nummer: 57910-98-2
UN: 3335
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR