1,2-Benzanthraquinone

Lieferant: Apollo Scientific
Achtung

Synonyme: Benz[a]anthracene-7,12-dione, Tetraphene-7,12-dione

OR54503-5G OR54503-1G OR54503-25G
APOSOR54503-5GEA 81.2 EUR
APOSOR54503-5G APOSOR54503-1G APOSOR54503-25G
1,2-Benzanthraquinone
1,2-Benzanthraquinone
Formel: C₁₈H₁₀O₂
Molecular Weight: 258,28 g/mol
Schmelzpunkt: 171 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003596
CAS-Nummer: 2498-66-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR