Ethyl hydrogen malonate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Mono-Ethyl malonate, 3-Ethoxy-3-oxopropanoic acid
OR54513-5G
OR54513-25G
APOSOR54513-5GEA
53.8
EUR
APOSOR54513-5G
APOSOR54513-25G
Ethyl hydrogen malonate
Ethyl hydrogen malonate
Formel:
C₅H₈O₄ Molecular Weight: 132,115 g/mol Siedepunkt: 208 °C Schmelzpunkt: –13 °C Dichte: 1,19 g/cm³ (25 °C) Flammpunkt: 113 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00020490 CAS-Nummer: 1071-46-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...