(S)-(-)-ɑ-Amino-γ-butyrolactone hydrochloride 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-α-Amino-γ-butyrolactone hydrochloride, L-(-)-Homoserine Lactone Hydrochloride, L-Homoserine lactone HCl, (S)-3-Aminodihydrofuran-2(3H)-one hydrochloride, L-Homoserine lactone hydrochloride HSI (S)-(-)-3-Aminotetrahydrofuran-2-one Hydrochloride, HIS, (S)-(-)-3-Aminotetrahydrofuran-2-one Hydrochloride

OR54538-100G OR54538-25G OR54538-5G
APOSOR54538-100GEA 1450 EUR
APOSOR54538-100G APOSOR54538-25G APOSOR54538-5G
(S)-(-)-ɑ-Amino-γ-butyrolactone hydrochloride 98%
(S)-(-)-alpha-Amino-gamma-butyrolactone hydrochloride
Formel: C₄H₇NO₂
Molecular Weight: 137.56 g/mol
Schmelzpunkt: 210…220 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00058172
CAS-Nummer: 2185-03-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR