2-Chlor-4-methylbenzonitril 98+%

Lieferant: Apollo Scientific
Achtung

OR5455-5G OR5455-1G OR5455-25G
APOSOR5455-5GEA 24.8 EUR
APOSOR5455-5G APOSOR5455-1G APOSOR5455-25G
2-Chlor-4-methylbenzonitril 98+%
2-Chlor-4-methylbenzonitril
Formel: C₈H₆CIN
Molecular Weight: 151.5900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094672
CAS-Nummer: 21423-84-7
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR