2,4-Diaminoanisol Dihydrochlorid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Methoxybenzene-1,3-diamine dihydrochloride

OR54554-25G OR54554-5G
APOSOR54554-25GEA 510 EUR
APOSOR54554-25G APOSOR54554-5G
2,4-Diaminoanisol Dihydrochlorid
2,4-Diaminoanisol Dihydrochlorid
Formel: C₇H₁₂Cl₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00054337
CAS-Nummer: 614-94-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR