Fmoc-L-phenylalanin 98%

Lieferant: Apollo Scientific

Synonyme: N-α-Fmoc-L-PhenylalanineN-(9-Fluorenylmethoxycarbonyl)-L-phenylalanineFmoc-Phe-OH

APOSOR54571-1KGEA 467 EUR
APOSOR54571-1KG APOSOR54571-100G APOSOR54571-500G
Fmoc-L-phenylalanin 98%
Fmoc-L-phenylalanin
Formel: C₂₄H₂₁NO₄
Molecular Weight: 387,4 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00037128
CAS-Nummer: 35661-40-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR