Ethyl-2-chloro-5-cyano-6-methylnicotinate

Lieferant: Apollo Scientific
Achtung

OR5458-1G OR5458-500MG
APOSOR5458-1GEA 33.5 EUR
APOSOR5458-1G APOSOR5458-500MG
Ethyl-2-chloro-5-cyano-6-methylnicotinate
Ethyl-2-chloro-5-cyano-6-methylnicotinate
Formel: C₁₀H₉ClN₂O₂
Molecular Weight: 224,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01565859
CAS-Nummer: 75894-43-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR