Ethyl-3-methylcrotonat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,3-Dimethylacrylic acid ethyl ester, Ethyl 3,3-Dimethylacrylate, Ethyl 3-methylbut-2-enoate, 3-Methyl-2-butenoic acid ethyl ester, 3-Methylcrotonic acid ethyl ester, Ethyl 3-Methyl-2-butenoate

OR54610-100G OR54610-25G
APOSOR54610-100GEA 115 EUR
APOSOR54610-100G APOSOR54610-25G
Ethyl-3-methylcrotonat 98%
Ethyl-3-methylcrotonat
Formel: C₇H₁₂O₂
Molecular Weight: 128.17 g/mol
Dichte: 0.92 g/cm³ (25 °C)
Flammpunkt: 43 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009187
CAS-Nummer: 638-10-8
UN: 3272
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR