4-Iodanilin 98%

Lieferant: Apollo Scientific
Achtung

OR5463-250G OR5463-50G OR5463-500G
APOSOR5463-250GEA 111 EUR
APOSOR5463-250G APOSOR5463-50G APOSOR5463-500G
4-Iodanilin 98%
4-Iodanilin
Formel: C₆H₆IN
Molecular Weight: 219,02 g/mol
Schmelzpunkt: 61…63 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007848
CAS-Nummer: 540-37-4
EINECS: 208-743-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR