H-Lys(boc)-OMe·HCl 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
H-Lys(Boc)-OMe·HCl, Nepsilon-Boc-L-lysine methyl ester hydrochloride, H-Lys(Boc)-OMe.HCl
OR54646-100G
OR54646-25G
APOSOR54646-100GEA
473
EUR
APOSOR54646-100G
APOSOR54646-25G
H-Lys(boc)-OMe·HCl 98%
H-Lys(boc)-OMe·HCl
Formel:
C₁₂H₂₅ClN₂O₄ Molecular Weight: 296,792 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00076959 CAS-Nummer: 2389-48-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...