4-Iodphenylboronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Jodbenzolboronsäure, 4-Iodbenzolboronsäure, (4-Iodphenyl)metaborsäure, (4-Jodphenyl)metaborsäure, Dihydroxy(4-jodphenyl)boran, Dihydroxy(4-iodphenyl)boran
OR5466-5G
OR5466-1G
APOSOR5466-5GEA
42.2
EUR
APOSOR5466-5G
APOSOR5466-1G
4-Iodphenylboronsäure 97%
4-Iodphenylboronsäure
Formel:
C₆H₆BIO₂ Molecular Weight: 247.83 g/mol Schmelzpunkt: 322…326 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01319014 CAS-Nummer: 5122-99-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...