5-Amino-2-brompyridin 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Brompyridin-5-amin, 2-Brom-5-pyridylamin
OR5471-1G
OR5471-25G
OR5471-5G
APOSOR5471-1GEA
14.7
EUR
APOSOR5471-1G
APOSOR5471-25G
APOSOR5471-5G
5-Amino-2-brompyridin 99%
5-Amino-2-brompyridin
Formel:
C₅H₅BrN₂ Molecular Weight: 173,01 g/mol Schmelzpunkt: 74…79 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01646116 CAS-Nummer: 13534-97-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...