Allyl-ɑ-D-galactopyranoside

Lieferant: Apollo Scientific

Synonyme: Ally a-D-galactopyranoside, Allyl α-D-Galactopyranoside, 2-Propen-1-yl α-D-Galactopyranoside, α-D-Galactose Monoallyl Ether

OR5475T-1G OR5475T-5G
APOSOR5475T-1GEA 139 EUR
APOSOR5475T-1G APOSOR5475T-5G
Allyl-ɑ-D-galactopyranoside
Allyl-alpha-D-galactopyranoside

A synthetic intermediated for oligosaccharide synthesis.

Formel: C₉H₁₆O₆
Molecular Weight: 220,22 g/mol
Schmelzpunkt: 138…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01320363
CAS-Nummer: 48149-72-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR