3-(Methylsulfonyl)benzoesäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR5476-100G
OR5476-25G
OR5476-5G
APOSOR5476-100GEA
640
EUR
APOSOR5476-100G
APOSOR5476-25G
APOSOR5476-5G
3-(Methylsulfonyl)benzoesäure 97%
3-(Methylsulphonyl)benzoic acid
Formel:
C₈H₈O₄S Molecular Weight: 200,21 g/mol Schmelzpunkt: 230…238 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00216488 CAS-Nummer: 5345-27-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...