3-(4,6-Dimethoxypyrimidin-2-yl)aniline 95+%

Lieferant: Apollo Scientific
Achtung

OR5478-1G OR5478-250MG
APOSOR5478-1GEA 0 EUR
APOSOR5478-1G APOSOR5478-250MG
3-(4,6-Dimethoxypyrimidin-2-yl)aniline 95+%
3-(4,6-Dimethoxypyrimidin-2-yl)aniline
Formel: C₁₂H₁₃N₃O₂
Schmelzpunkt: 100…101 °C
MDL: MFCD01319521
CAS-Nummer: 387350-84-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR