7-Aminoimidazo[1,2-a]pyridin

Lieferant: Apollo Scientific
Achtung

Synonyme: Imidazo[1,2-a]pyridin-7-amine

OR55000-250MG OR55000-1G
APOSOR55000-250MGEA 72.5 EUR
APOSOR55000-250MG APOSOR55000-1G
7-Aminoimidazo[1,2-a]pyridin
7-Aminoimidazo[1,2-a]pyridin
Formel: C₇H₇N₃
Molecular Weight: 133.15 g/mol
Schmelzpunkt: 139…141 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11052644
CAS-Nummer: 421595-81-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR