2-Methyl-2-(pyridin-4-yl)propan-1-amine 95+%

Lieferant: Apollo Scientific

Synonyme: 4-(1-Amino-2-methylprop-2-yl)pyridine

OR55002-1G OR55002-5G
APOSOR55002-1GEA 592 EUR
APOSOR55002-1G APOSOR55002-5G
2-Methyl-2-(pyridin-4-yl)propan-1-amine 95+%
2-Methyl-2-(pyridin-4-yl)propan-1-amine
Formel: C₉H₁₄N₂
Molecular Weight: 150,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11898942
CAS-Nummer: 1060815-29-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR