6-Hydroxy-2H-chromene-3-carboxaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Hydroxy-4H-chromene-3-carbaldehyde
OR55042-1G
OR55042-250MG
APOSOR55042-1GEA
55.1
EUR
APOSOR55042-1G
APOSOR55042-250MG
6-Hydroxy-2H-chromene-3-carboxaldehyde
6-Hydroxy-2H-chromene-3-carboxaldehyde
Formel:
C₁₀H₈O₃ Molecular Weight: 176,17 g/mol Schmelzpunkt: 164…169 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01076587 CAS-Nummer: 134822-76-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...