(S)-(+)-2-Chloro-1-phenylethanol

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-2-Chloro-1-phenylethanol, (S)-(+)-alpha-(Chloromethyl)benzyl alcohol

OR55062-1G OR55062-250MG
APOSOR55062-1GEA 79.9 EUR
APOSOR55062-1G APOSOR55062-250MG
(S)-(+)-2-Chloro-1-phenylethanol
(S)-(+)-2-Chloro-1-phenylethanol
Formel: C₈H₉ClO
Molecular Weight: 156,61 g/mol
Dichte: 1,185 g/cm³ (25 °C)
Flammpunkt: 113 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00077692
CAS-Nummer: 70111-05-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR