Diethyl furan-2,5-dicarboxylate 99%
Lieferant: Apollo Scientific
|
Achtung
|
OR55064-5G
OR55064-25G
APOSOR55064-5GEA
26.1
EUR
APOSOR55064-5G
APOSOR55064-25G
Diethyl furan-2,5-dicarboxylate 99%
Diethyl furan-2,5-dicarboxylate
Formel:
C₁₀H₁₂O₅ Schmelzpunkt: 45…47 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00449457 CAS-Nummer: 53662-83-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...