1-Benzyl-2-pyrrolidinon 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Benzylpyrrolidin-2-one
OR55077-100G
OR55077-500G
APOSOR55077-100GEA
168
EUR
APOSOR55077-100G
APOSOR55077-500G
1-Benzyl-2-pyrrolidinon 99%
1-Benzyl-2-pyrrolidinon
Formel:
C₁₁H₁₃NO Molecular Weight: 175,23 g/mol Dichte: 1,095 g/cm³ (25 °C) Flammpunkt: 113 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003195 CAS-Nummer: 5291-77-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...