2-(Dicyclohexylphosphino)-2'-methylbiphenyl
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Dicyclohexyl(2'-methyl-[1,1'-biphenyl]-2-yl)phosphine, 2-Dicyclohexylphosphino-2'-methyl-)-1,1'-biphenyl, MePhos, 2-Dicyclohexylphosphino-2'-methylbiphenyl
OR55088-100G
OR55088-25G
APOSOR55088-100GEA
244
EUR
APOSOR55088-100G
APOSOR55088-25G
2-(Dicyclohexylphosphino)-2'-methylbiphenyl
2-(Dicyclohexylphosphino)-2'-methylbiphenyl
Formel:
C₂₅H₃₃P Molecular Weight: 364,513 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD03094577 CAS-Nummer: 251320-86-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...