4-Formyl-1-methylpyridinium benzenesulphonate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Formyl-1-methylpyridin-1-ium benzenesulphonate, N-Methylpyridinium-4-carboxaldehyde benzenesulphonate
OR55120-1G
OR55120-5G
APOSOR55120-1GEA
125
EUR
APOSOR55120-1G
APOSOR55120-5G
4-Formyl-1-methylpyridinium benzenesulphonate
4-Formyl-1-methylpyridinium benzenesulphonate
Formel:
C₁₃H₁₃NO₄S Molecular Weight: 279,31 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043164 CAS-Nummer: 82228-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...