D-Erythronolactone 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3R,4R)-3,4-Dihydroxydihydrofuran-2(3H)-one, D-Erythronic gamma-Lactone, D-Erythronolactone, D-Erythrono-1,4-lactone, (3R,4R)-Dihydroxydihydro-2(3H)-furanone
OR55152-25G
APOSOR55152-25GEA
95.7
EUR
APOSOR55152-25G
D-Erythronolactone 98%
D-Erythronolactone
Formel:
C₄H₆O₄ Molecular Weight: 118,09 g/mol Schmelzpunkt: 103 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00077763 CAS-Nummer: 15667-21-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...