(2R,4R)-4-Methylpiperidine-2-carboxylic acid
Lieferant: Apollo Scientific
OR55190-1G
OR55190-5G
APOSOR55190-1GEA
79.9
EUR
APOSOR55190-1G
APOSOR55190-5G
(2R,4R)-4-Methylpiperidine-2-carboxylic acid
(2R,4R)-4-Methylpiperidine-2-carboxylic acid
Formel:
C₇H₁₃NO₂ Molecular Weight: 143.1800 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03839858 CAS-Nummer: 74892-81-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...