Boc-D-trans-Hyp-OMe
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2R,4S)-1-tert-Butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate
OR55193-25G
OR55193-5G
APOSOR55193-25GEA
192
EUR
APOSOR55193-25G
APOSOR55193-5G
Boc-D-trans-Hyp-OMe
Boc-D-trans-Hyp-OMe
Formel:
C₁₁H₁₉NO₅ Molecular Weight: 245,3 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09954932 CAS-Nummer: 135042-17-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...