trans-4-Amino-1-methylcyclohexanol 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR55239-100MG
OR55239-1G
OR55239-250MG
APOSOR55239-100MGEA
109
EUR
APOSOR55239-100MG
APOSOR55239-1G
APOSOR55239-250MG
trans-4-Amino-1-methylcyclohexanol 97%
trans-4-Amino-1-methylcyclohexanol
Formel:
C₇H₁₅NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD18632623 CAS-Nummer: 177908-37-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...