2-Amino-4-chlor-5-nitrophenol

Lieferant: Apollo Scientific
Achtung

OR55270-100G OR55270-25G
APOSOR55270-100GEA 452 EUR
APOSOR55270-100G APOSOR55270-25G
2-Amino-4-chlor-5-nitrophenol
2-Amino-4-chlor-5-nitrophenol
Formel: C₆H₅ClN₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00010300
CAS-Nummer: 6358-07-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR