1-(1-Boc-piperidino)-4-brom-1H-pyrazol
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl 4-(4-bromo-1H-pyrazol-1-yl)piperidine-1-carboxylate, 1-(4-Boc-1-piperidinyl)-4-bromopyrazole
OR55287-25G
OR55287-5G
APOSOR55287-25GEA
203
EUR
APOSOR55287-25G
APOSOR55287-5G
1-(1-Boc-piperidino)-4-brom-1H-pyrazol
1-(1-Boc-piperidino)-4-brom-1H-pyrazol
Formel:
C₁₃H₂₀BrN₃O₂ Molecular Weight: 330.23 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09831980 CAS-Nummer: 877399-50-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...